siRNA Details
Gene Symbol: Rfx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107694.1 | NP_001101164.1 | 4, 5 | 277 | ||
XM_006232876.2 | XP_006232938.1 | 3, 4 | 1775 | |||
XM_006232878.2 | XP_006232940.1 | 3, 4 | 2084 | |||
XM_008761300.1 | XP_008759522.1 | 2, 3 | 526 | |||
XM_008761301.1 | XP_008759523.1 | 3, 4 | 2030 |
