siRNA Details
| Gene Symbol: Hfe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012080.1 | NP_001012080.1 | 3 | 705 | ||
| GenBank mRNA | BC089203.1 | - | 3 | 705 | 7191112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hfe2| Gene Symbol: Hfe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012080.1 | NP_001012080.1 | 3 | 705 | ||
| GenBank mRNA | BC089203.1 | - | 3 | 705 | 7191112 | |