siRNA Details
| Gene Symbol: Polr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012081.1 | NP_001012081.1 | 4 | 790 | ||
| XM_006232967.1 | XP_006233029.1 | 4 | 997 | |||
| XM_008761310.1 | XP_008759532.1 | 4 | 720 | |||
| XM_008761311.1 | XP_008759533.1 | 4 | 997 | |||
| GenBank mRNA | BC083667.1 | - | 4 | 790 | 7135563 | |

Polr3c
Genomic Map