siRNA Details
| Gene Symbol: Prcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107700.1 | NP_001101170.1 | 6 | 1448 | ||
| GenBank mRNA | BC166708.1 | - | 6 | 1448 | 5621845 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prcc| Gene Symbol: Prcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107700.1 | NP_001101170.1 | 6 | 1448 | ||
| GenBank mRNA | BC166708.1 | - | 6 | 1448 | 5621845 | |