siRNA Details
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107700.1 | NP_001101170.1 | 4 | 1100 | ||
GenBank mRNA | BC166708.1 | - | 4 | 1100 | 5621845 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107700.1 | NP_001101170.1 | 4 | 1100 | ||
GenBank mRNA | BC166708.1 | - | 4 | 1100 | 5621845 |