siRNA Details
| Gene Symbol: Sec22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025686.1 | NP_001020857.1 | 4 | 521 | ||
| GenBank mRNA | BC098725.1 | - | 4 | 521 | 7378284 | |
| FQ211503.1 | - | 546 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec22b| Gene Symbol: Sec22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025686.1 | NP_001020857.1 | 4 | 521 | ||
| GenBank mRNA | BC098725.1 | - | 4 | 521 | 7378284 | |
| FQ211503.1 | - | 546 | ||||