siRNA Details
| Gene Symbol: Mov10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107711.1 | NP_001101181.1 | 17 | 2782 | ||
| XM_006233085.1 | XP_006233147.1 | 17 | 2670 | |||
| XM_008761395.1 | XP_008759617.1 | 17 | 3530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mov10| Gene Symbol: Mov10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107711.1 | NP_001101181.1 | 17 | 2782 | ||
| XM_006233085.1 | XP_006233147.1 | 17 | 2670 | |||
| XM_008761395.1 | XP_008759617.1 | 17 | 3530 | |||