siRNA Details
| Gene Symbol: Lrif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127485.1 | NP_001120957.1 | 2, 3 | 1701 | ||
| XM_006233147.2 | XP_006233209.1 | 1, 2 | 176 | |||
| XM_006233149.2 | XP_006233211.1 | 1, 2 | 1529 | |||
| XM_008761400.1 | XP_008759622.1 | 2, 3 | 1535 | |||
| GenBank mRNA | BC099834.1 | - | 1, 2 | 118 | ||
| BC158797.1 | - | 1, 2 | 72 | |||

Lrif1
Genomic Map