siRNA Details
| Gene Symbol: Lrif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127485.1 | NP_001120957.1 | 2 | 1045 | ||
| XM_008761400.1 | XP_008759622.1 | 2 | 879 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrif1| Gene Symbol: Lrif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127485.1 | NP_001120957.1 | 2 | 1045 | ||
| XM_008761400.1 | XP_008759622.1 | 2 | 879 | |||