siRNA Details
| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761474.1 | XP_008759696.1 | 17 | 1971 | ||
| GenBank mRNA | FQ210266.1 | - | 17 | 1908 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frrs1| Gene Symbol: Frrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761474.1 | XP_008759696.1 | 17 | 1971 | ||
| GenBank mRNA | FQ210266.1 | - | 17 | 1908 | ||