siRNA Details
| Gene Symbol: Sec24d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749389.3 | XP_003749437.1 | 15, 16 | 2124 | ||
| XM_006233272.2 | XP_006233334.1 | 15, 16 | 2121 | |||
| GenBank mRNA | BC168946.1 | - | 8, 9 | 1034 | 8366565 | |

Sec24d
Genomic Map