siRNA Details
| Gene Symbol: Sec24d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749389.3 | XP_003749437.1 | 14 | 1891 | ||
| XM_006233272.2 | XP_006233334.1 | 14 | 1888 | |||
| GenBank mRNA | BC168946.1 | - | 7 | 801 | 8366565 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec24d| Gene Symbol: Sec24d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749389.3 | XP_003749437.1 | 14 | 1891 | ||
| XM_006233272.2 | XP_006233334.1 | 14 | 1888 | |||
| GenBank mRNA | BC168946.1 | - | 7 | 801 | 8366565 | |