siRNA Details
| Gene Symbol: Ppa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135871.1 | NP_001129343.1 | 5, 6 | 433 | ||
| XM_006233327.2 | XP_006233389.1 | 5, 6 | 2030 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppa2| Gene Symbol: Ppa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135871.1 | NP_001129343.1 | 5, 6 | 433 | ||
| XM_006233327.2 | XP_006233389.1 | 5, 6 | 2030 | |||