siRNA Details
| Gene Symbol: Mttp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107727.1 | NP_001101197.1 | 10 | 1370 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mttp| Gene Symbol: Mttp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107727.1 | NP_001101197.1 | 10 | 1370 | ||