siRNA Details
| Gene Symbol: Fpgt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199494.1 | NP_955788.1 | 4 | 1334 | ||
| GenBank mRNA | AJ276066.1 | - | 4 | 1369 | ||
| BC168244.1 | - | 4 | 1314 | 5600080 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpgt| Gene Symbol: Fpgt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199494.1 | NP_955788.1 | 4 | 1334 | ||
| GenBank mRNA | AJ276066.1 | - | 4 | 1369 | ||
| BC168244.1 | - | 4 | 1314 | 5600080 | ||