siRNA Details
| Gene Symbol: Itgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004263.1 | NP_001004263.1 | 8, 9 | 1081 | ||
| XM_006234232.1 | XP_006234294.1 | 10, 11 | 1424 | |||
| XM_006234233.2 | XP_006234295.1 | 8, 9 | 1330 | |||
| XM_008761845.1 | XP_008760067.1 | 10, 11 | 1251 | |||
| XM_008761846.1 | XP_008760068.1 | 8, 9 | 1228 | |||
| GenBank mRNA | BC079069.1 | - | 8, 9 | 1081 | 7121642 | |

Itgb6
Genomic Map