siRNA Details
Gene Symbol: Xirp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201989.1 | NP_973718.1 | 7 | 7689 | ||
GenBank mRNA | AF466694.1 | - | 7 | 7689 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Xirp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201989.1 | NP_973718.1 | 7 | 7689 | ||
GenBank mRNA | AF466694.1 | - | 7 | 7689 |