siRNA Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024260.1 | NP_001019431.1 | 1, 2 | 73 | ||
| GenBank mRNA | BC088446.1 | - | 1, 2 | 73 | 7126459 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nostrin| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024260.1 | NP_001019431.1 | 1, 2 | 73 | ||
| GenBank mRNA | BC088446.1 | - | 1, 2 | 73 | 7126459 | |