siRNA Details
| Gene Symbol: Tlk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107734.1 | NP_001101204.1 | 588 | |||
| XM_008761896.1 | XP_008760118.1 | 6, 7 | 639 | |||
| XM_008761897.1 | XP_008760119.1 | 1871 | ||||
| XM_008761898.1 | XP_008760120.1 | 6, 7 | 800 | |||
| XM_008761899.1 | XP_008760121.1 | 421 | ||||
| Gene Symbol: LOC103690017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234342.2 | XP_006234404.2 | 6, 7 | 421 | ||
| XM_008761952.1 | XP_008760174.1 | 6, 7 | 1871 | |||

Tlk1
Genomic Map