siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107735.1 | NP_001101205.1 | 14, 15 | 1877 | ||
| XM_008761901.1 | XP_008760123.1 | 12, 13 | 1371 | |||
| XM_008761904.1 | XP_008760126.1 | 11, 12 | 1278 | |||
| XM_008761906.1 | XP_008760128.1 | 10, 11 | 1203 | |||

Osbpl6
Genomic Map