siRNA Details
| Gene Symbol: Osbpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107735.1 | NP_001101205.1 | 20 | 2644 | ||
| XM_008761900.1 | XP_008760122.1 | 19 | 2162 | |||
| XM_008761901.1 | XP_008760123.1 | 18 | 2138 | |||
| XM_008761902.1 | XP_008760124.1 | 18 | 2087 | |||
| XM_008761903.1 | XP_008760125.1 | 18 | 2069 | |||
| XM_008761904.1 | XP_008760126.1 | 17 | 2045 | |||
| XM_008761905.1 | XP_008760127.1 | 17 | 1994 | |||
| XM_008761906.1 | XP_008760128.1 | 16 | 1970 | |||

Osbpl6
Genomic Map