siRNA Details
| Gene Symbol: Lrrc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122975.1 | NP_001116447.1 | 1 | 716 | ||
| XM_006234472.2 | XP_006234534.1 | 2 | 1828 | |||
| GenBank mRNA | BC099134.1 | - | 1 | 716 | 7454777 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc55| Gene Symbol: Lrrc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122975.1 | NP_001116447.1 | 1 | 716 | ||
| XM_006234472.2 | XP_006234534.1 | 2 | 1828 | |||
| GenBank mRNA | BC099134.1 | - | 1 | 716 | 7454777 | |