siRNA Details
| Gene Symbol: Ddb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271346.1 | NP_001258275.1 | 1 | 275 | ||
| XM_006234498.2 | XP_006234560.1 | 1 | 307 | |||
| XM_006234499.2 | XP_006234561.1 | 1 | 308 | |||
| GenBank mRNA | FQ228020.1 | - | 1 | 275 | ||

Ddb2
Genomic Map