siRNA Details
| Gene Symbol: LOC100362202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749532.3 | XP_003749580.1 | 5 | 2220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100362202| Gene Symbol: LOC100362202 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749532.3 | XP_003749580.1 | 5 | 2220 | ||