siRNA Details
| Gene Symbol: Ttc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107752.2 | NP_001101222.2 | 3, 4 | 423 | ||
| XM_006234582.2 | XP_006234644.1 | 3, 4 | 452 | |||
| GenBank mRNA | BC168714.1 | - | 3, 4 | 423 | 9025859 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc17| Gene Symbol: Ttc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107752.2 | NP_001101222.2 | 3, 4 | 423 | ||
| XM_006234582.2 | XP_006234644.1 | 3, 4 | 452 | |||
| GenBank mRNA | BC168714.1 | - | 3, 4 | 423 | 9025859 | |