siRNA Details
| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107754.2 | NP_001101224.1 | 7 | 1040 | ||
| GenBank mRNA | BC168921.1 | - | 7 | 1040 | 9024408 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf6| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107754.2 | NP_001101224.1 | 7 | 1040 | ||
| GenBank mRNA | BC168921.1 | - | 7 | 1040 | 9024408 | |