siRNA Details
| Gene Symbol: Arl14ep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014045.1 | NP_001014067.1 | 2 | 245 | ||
| XM_008762092.1 | XP_008760314.1 | 2 | 1626 | |||
| GenBank mRNA | BC089920.1 | - | 2 | 245 | 7306710 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl14ep| Gene Symbol: Arl14ep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014045.1 | NP_001014067.1 | 2 | 245 | ||
| XM_008762092.1 | XP_008760314.1 | 2 | 1626 | |||
| GenBank mRNA | BC089920.1 | - | 2 | 245 | 7306710 | |