siRNA Details
| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107759.2 | NP_001101229.2 | 8 | 4110 | ||
| XM_006234756.2 | XP_006234818.1 | 8 | 4124 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Disp2| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107759.2 | NP_001101229.2 | 8 | 4110 | ||
| XM_006234756.2 | XP_006234818.1 | 8 | 4124 | |||