siRNA Details
| Gene Symbol: LOC100911166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749554.3 | XP_003749602.1 | 1597 | |||
| Gene Symbol: Rpusd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135845.1 | NP_001129317.1 | 3 | 1602 | ||

LOC100911166
Genomic Map