siRNA Details
Gene Symbol: LOC100911166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749554.3 | XP_003749602.1 | 759 |
Gene Symbol: Rpusd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135845.1 | NP_001129317.1 | 2 | 764 |
