siRNA Details
| Gene Symbol: Rmdn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014046.2 | NP_001014068.1 | 6 | 987 | ||
| GenBank mRNA | BC082081.1 | - | 6 | 949 | 7133344 | |
| Gene Symbol: LOC100911313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762136.1 | XP_008760358.1 | 3 | 667 | ||

Rmdn3
Genomic Map