siRNA Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107760.1 | NP_001101230.1 | 9 | 2012 | ||
| XM_006234765.2 | XP_006234827.1 | 9 | 2023 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dll4| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107760.1 | NP_001101230.1 | 9 | 2012 | ||
| XM_006234765.2 | XP_006234827.1 | 9 | 2023 | |||