siRNA Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107761.1 | NP_001101231.1 | 12 | 2470 | ||
| XM_006234766.2 | XP_006234828.1 | 12 | 2231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exd1| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107761.1 | NP_001101231.1 | 12 | 2470 | ||
| XM_006234766.2 | XP_006234828.1 | 12 | 2231 | |||