siRNA Details
| Gene Symbol: Nusap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107762.1 | NP_001101232.1 | 7 | 962 | ||
| XM_006234768.2 | XP_006234830.1 | 7 | 796 | |||
| XM_006234769.2 | XP_006234831.1 | 7 | 793 | |||
| XM_006234770.2 | XP_006234832.1 | 6 | 649 | |||
| XM_008762101.1 | XP_008760323.1 | 6 | 706 | |||
| XM_008762102.1 | XP_008760324.1 | 6 | 703 | |||
| GenBank mRNA | BC161947.1 | - | 7 | 741 | 9025380 | |

Nusap1
Genomic Map