siRNA Details
| Gene Symbol: Nusap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107762.1 | NP_001101232.1 | 9 | 1259 | ||
| XM_006234768.2 | XP_006234830.1 | 10 | 1210 | |||
| XM_006234769.2 | XP_006234831.1 | 10 | 1207 | |||
| XM_006234770.2 | XP_006234832.1 | 9 | 1063 | |||
| XM_008762101.1 | XP_008760323.1 | 9 | 1120 | |||
| XM_008762102.1 | XP_008760324.1 | 9 | 1117 | |||
| GenBank mRNA | BC161947.1 | - | 10 | 1155 | 9025380 | |

Nusap1
Genomic Map