siRNA Details
| Gene Symbol: Pla2g4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107764.2 | NP_001101234.2 | 18 | 1938 | ||
| XM_006234775.1 | XP_006234837.1 | 17 | 1870 | |||
| XM_008762103.1 | XP_008760325.1 | 17 | 2511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g4b| Gene Symbol: Pla2g4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107764.2 | NP_001101234.2 | 18 | 1938 | ||
| XM_006234775.1 | XP_006234837.1 | 17 | 1870 | |||
| XM_008762103.1 | XP_008760325.1 | 17 | 2511 | |||