siRNA Details
Gene Symbol: Tmem62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230497.8 | XP_230497.4 | 10 | 2016 | ||
GenBank mRNA | FQ210040.1 | - | 10 | 1420 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230497.8 | XP_230497.4 | 10 | 2016 | ||
GenBank mRNA | FQ210040.1 | - | 10 | 1420 |