siRNA Details
Gene Symbol: Frmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762211.1 | XP_008760433.1 | 10 | 793 | ||
XM_008762212.1 | XP_008760434.1 | 10 | 808 | |||
XM_008762213.1 | XP_008760435.1 | 10 | 793 | |||
XM_008762214.1 | XP_008760436.1 | 10 | 836 | |||
XM_008762215.1 | XP_008760437.1 | 10 | 836 |
