siRNA Details
| Gene Symbol: Sema6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107768.1 | NP_001101238.1 | 2 | 530 | ||
| XM_008762171.1 | XP_008760393.1 | 1 | 3903 | |||
| XM_008762172.1 | XP_008760394.1 | 1 | 3903 | |||
| XM_008762173.1 | XP_008760395.1 | 1 | 3903 | |||
| XM_008762174.1 | XP_008760396.1 | 1 | 3903 | |||
| XM_008762175.1 | XP_008760397.1 | 1 | 3903 | |||

Sema6d
Genomic Map