siRNA Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107770.1 | NP_001101240.1 | 7 | 1019 | ||
| XM_006234920.2 | XP_006234982.1 | 7 | 880 | |||
| GenBank mRNA | FQ210986.1 | - | 5 | 465 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sppl2a| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107770.1 | NP_001101240.1 | 7 | 1019 | ||
| XM_006234920.2 | XP_006234982.1 | 7 | 880 | |||
| GenBank mRNA | FQ210986.1 | - | 5 | 465 | ||