siRNA Details
Gene Symbol: Stk35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107773.1 | NP_001101243.1 | 3 | 2651 | ||
XM_006234974.2 | XP_006235036.1 | 3 | 1599 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stk35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107773.1 | NP_001101243.1 | 3 | 2651 | ||
XM_006234974.2 | XP_006235036.1 | 3 | 1599 |