siRNA Details
| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107775.1 | NP_001101245.1 | 11 | 1486 | ||
| GenBank mRNA | FQ225785.1 | - | 11 | 1480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc4a11| Gene Symbol: Slc4a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107775.1 | NP_001101245.1 | 11 | 1486 | ||
| GenBank mRNA | FQ225785.1 | - | 11 | 1480 | ||