siRNA Details
| Gene Symbol: Kif16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107783.2 | NP_001101253.2 | 21, 22 | 3475 | ||
| XM_006235108.2 | XP_006235170.1 | 22, 23 | 3661 | |||
| XM_006235109.2 | XP_006235171.1 | 21, 22 | 3628 | |||
| XM_006235112.2 | XP_006235174.1 | 22, 23 | 3643 | |||
| XM_006235113.2 | XP_006235175.1 | 22, 23 | 3643 | |||
| XM_008762263.1 | XP_008760485.1 | 23, 24 | 3549 | |||
| XM_008762264.1 | XP_008760486.1 | 22, 23 | 3473 | |||
| XM_008762265.1 | XP_008760487.1 | 22, 23 | 4237 | |||
| XM_008762266.1 | XP_008760488.1 | 21, 22 | 3546 | |||
| XM_008762267.1 | XP_008760489.1 | 22, 23 | 3643 | |||
| XM_008762268.1 | XP_008760490.1 | 22, 23 | 3643 | |||

Kif16b
Genomic Map