siRNA Details
| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762285.1 | XP_008760507.1 | 10 | 996 | ||
| XM_008762286.1 | XP_008760508.1 | 10 | 984 | |||
| XM_008762287.1 | XP_008760509.1 | 10 | 1032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap61| Gene Symbol: Cfap61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762285.1 | XP_008760507.1 | 10 | 996 | ||
| XM_008762286.1 | XP_008760508.1 | 10 | 984 | |||
| XM_008762287.1 | XP_008760509.1 | 10 | 1032 | |||