siRNA Details
| Gene Symbol: Tpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107790.1 | NP_001101260.1 | 11, 12 | 1799 | ||
| XM_006235270.2 | XP_006235332.1 | 12, 13 | 1702 | |||
| XM_006235272.2 | XP_006235334.1 | 12, 13 | 1654 | |||
| XM_006235273.2 | XP_006235335.1 | 11, 12 | 1584 | |||
| XM_006235274.2 | XP_006235336.1 | 12, 13 | 1702 | |||

Tpx2
Genomic Map