siRNA Details
Gene Symbol: Tpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107790.1 | NP_001101260.1 | 9, 10 | 1448 | ||
XM_006235270.2 | XP_006235332.1 | 10, 11 | 1351 | |||
XM_006235272.2 | XP_006235334.1 | 10, 11 | 1303 | |||
XM_006235273.2 | XP_006235335.1 | 9, 10 | 1233 | |||
XM_006235274.2 | XP_006235336.1 | 10, 11 | 1351 |
