siRNA Details
| Gene Symbol: Xkr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012092.1 | NP_001012092.1 | 3 | 1060 | ||
| XM_008762334.1 | XP_008760556.1 | 4 | 2784 | |||
| GenBank mRNA | AY534261.1 | - | 3 | 1060 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr7| Gene Symbol: Xkr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012092.1 | NP_001012092.1 | 3 | 1060 | ||
| XM_008762334.1 | XP_008760556.1 | 4 | 2784 | |||
| GenBank mRNA | AY534261.1 | - | 3 | 1060 | ||