siRNA Details
| Gene Symbol: Itch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005887.1 | NP_001005887.1 | 10, 11 | 1186 | ||
| XM_008762335.1 | XP_008760557.1 | 12, 13 | 1414 | |||
| XM_008762336.1 | XP_008760558.1 | 12, 13 | 1339 | |||
| XM_008762337.1 | XP_008760559.1 | 9, 10 | 1091 | |||
| GenBank mRNA | AY600518.1 | - | 10, 11 | 1186 | ||

Itch
Genomic Map