siRNA Details
| Gene Symbol: Phf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107795.1 | NP_001101265.1 | 3 | 378 | ||
| XM_006235404.2 | XP_006235466.1 | 4 | 454 | |||
| XM_008762342.1 | XP_008760564.1 | 4 | 465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf20| Gene Symbol: Phf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107795.1 | NP_001101265.1 | 3 | 378 | ||
| XM_006235404.2 | XP_006235466.1 | 4 | 454 | |||
| XM_008762342.1 | XP_008760564.1 | 4 | 465 | |||