siRNA Details
| Gene Symbol: Rprd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098727.1 | NP_001092197.1 | 4, 5 | 896 | ||
| XM_006235412.1 | XP_006235474.1 | 4, 5 | 899 | |||
| GenBank mRNA | BC168699.1 | - | 4, 5 | 901 | 8362231 | |
| CK365545.1 | - | 2, 3 | 144 | |||

Rprd1b
Genomic Map